Assay Details
Target Gene Details
Entrez Gene ID: | 58526 |
Gene Name: | MID1 interacting protein 1 |
Gene Aliases: |
G12-like, MIG12, S14R, STRAIT11499, THRSPL |
Location: |
Chr.X:38801432-38806532 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MID1IP1 | NM_001098790.1 | 3 | 457 | NP_001092260.1 |
| NM_001098791.1 | 2 | 353 | NP_001092261.1 | |
| NM_021242.5 | 3 | 2232 | NP_067065.1 | |
| AJ272057.1 | 2 | 492 | CAB89113.1 | |
| AK001248.1 | 2 | 439 | BAA91580.1 | |
| BC008908.2 | 2 | 891 | AAH08908.1 | |
| BC019332.1 | 3 | 435 | AAH19332.1 | |
| CR457220.1 | CAG33501.1 | |||
| DA138791.1 | 2 | 353 |
More Information
Set Membership: |
|

Genomic Map