Assay Details
Target Gene Details
Entrez Gene ID: | 23097 |
Gene Name: | cyclin dependent kinase 19 |
Gene Aliases: |
CDC2L6, CDK11, bA346C16.3 |
Location: |
Chr.6:110609978-110816531 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK19 | NM_001300960.1 | 12 | 4632 | NP_001287889.1 |
| NM_001300963.1 | 14 | 4580 | NP_001287892.1 | |
| NM_001300964.1 | 13 | 4534 | NP_001287893.1 | |
| NM_015076.4 | 13 | 4764 | NP_055891.1 | |
| XM_005266871.3 | 10 | 4347 | XP_005266928.1 | |
| XM_011535630.2 | 13 | 5110 | XP_011533932.1 | |
| XM_011535631.2 | 12 | 4982 | XP_011533933.1 | |
| XM_017010587.1 | 14 | 4990 | XP_016866076.1 | |
| XM_017010588.1 | 11 | 4456 | XP_016866077.1 | |
| AB028951.2 | 13 | 4581 | BAA82980.2 | |
| AL122055.1 | 10 | 4219 | CAB59186.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830770 | Chr.6:110513882 - 110702928 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map