Assay Details
Target Gene Details
Entrez Gene ID: | 284467 |
Gene Name: | family with sequence similarity 19 member A3, C-C motif chemokine like |
Gene Aliases: |
TAFA-3, TAFA3 |
Location: |
Chr.1:112719003-112727235 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A3 | NM_001004440.1 | NP_001004440.1 | ||
| NM_182759.2 | NP_877436.1 | |||
| XM_005270766.3 | XP_005270823.1 | |||
| XM_006710582.3 | XP_006710645.1 | |||
| AY325116.1 | AAP92408.1 | |||
| AY325119.1 | AAP92411.1 | |||
| BC112268.1 | AAI12269.1 | |||
| BC117479.1 | ||||
| BC144043.1 | ||||
| BQ638101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818378 | Chr.1:112721089 - 112892196 on Build GRCh38 | Gain |
|
| nsv547566 | Chr.1:112705743 - 112759035 on Build GRCh38 | Gain |
|
| nsv998794 | Chr.1:112704008 - 112767796 on Build GRCh38 | Gain |
|
| esv27598 | Chr.1:112703696 - 112808153 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map