Assay Details
Target Gene Details
Entrez Gene ID: | 8872 |
Gene Name: | cell division cycle 123 |
Gene Aliases: |
C10orf7, D123 |
Location: |
Chr.10:12195962-12250590 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC123 | NM_006023.2 | 1 | 195 | NP_006014.2 |
| XM_005252638.3 | XP_005252695.1 | |||
| AK225634.1 | 1 | 161 | ||
| AK308790.1 | ||||
| BC009598.1 | AAH09598.1 | |||
| BP367374.1 | 1 | 195 | ||
| D14878.1 | 1 | 191 | BAA03593.1 | |
| U27112.1 | AAC34738.1 |
Target Gene Details
Entrez Gene ID: | 11164 |
Gene Name: | nudix hydrolase 5 |
Gene Aliases: |
YSA1, YSA1H, YSAH1 |
Location: |
Chr.10:12165328-12196144 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT5 | NM_001321647.1 | NP_001308576.1 | ||
| NM_001321648.1 | NP_001308577.1 | |||
| NM_014142.3 | NP_054861.2 | |||
| AK314144.1 | ||||
| BU602000.1 |
More Information
Set Membership: |
|

Genomic Map