Assay Details
Target Gene Details
Entrez Gene ID: | 7374 |
Gene Name: | uracil DNA glycosylase |
Gene Aliases: |
DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2 |
Location: |
Chr.12:109097594-109110993 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNG | NM_003362.3 | 6 | 1272 | NP_003353.1 |
| NM_080911.2 | 7 | 1176 | NP_550433.1 | |
| AK291341.1 | 6 | 1196 | ||
| AK299416.1 | 7 | 1116 | ||
| BC015205.1 | 6 | 1155 | AAH15205.1 | |
| BC050634.2 | 6 | 1166 | AAH50634.1 | |
| CR749342.1 | 5 | 1142 | CAH18195.1 | |
| X15653.1 | 6 | 1169 | CAA33679.1 | |
| Y09008.1 | 7 | 1160 | CAA70211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1553n100 | Chr.12:109064624 - 109159352 on Build GRCh38 | Gain |
|
| esv2422361 | Chr.12:108792678 - 109118360 on Build GRCh38 | Duplication |
|
| dgv1554n100 | Chr.12:109104860 - 109232663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map