Assay Details
Target Gene Details
Entrez Gene ID: | 57125 |
Gene Name: | plexin domain containing 1 |
Gene Aliases: |
TEM3, TEM7 |
Location: |
Chr.17:39063303-39151649 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLXDC1 | NM_020405.4 | NP_065138.2 | ||
| AF279144.2 | AAG00869.2 | |||
| AF378753.1 | AAL11990.1 | |||
| AK093589.1 | ||||
| AK127539.1 | BAC87025.1 | |||
| AK303699.1 | ||||
| AK312999.1 | ||||
| AY704670.1 | AAV85657.1 | |||
| AY704671.1 | AAV85658.1 | |||
| AY704672.1 | AAV85659.1 | |||
| BC036059.1 | AAH36059.1 | |||
| KU257674.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833440 | Chr.17:39033205 - 39210437 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map