Assay Details
Target Gene Details
Entrez Gene ID: | 28992 |
Gene Name: | MACRO domain containing 1 |
Gene Aliases: |
LRP16 |
Location: |
Chr.11:63998554-64166652 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MACROD1 | NM_014067.3 | NP_054786.2 | ||
| XM_005273939.4 | XP_005273996.1 | |||
| XM_011544970.2 | XP_011543272.1 | |||
| AF202922.2 | AAF15294.2 | |||
| BC000270.1 | AAH00270.2 | |||
| BC003188.1 | AAH03188.1 | |||
| BC007297.1 | AAH07297.1 | |||
| BC008316.1 | AAH08316.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| nsv1042075 | Chr.11:63964982 - 64045692 on Build GRCh38 | Loss |
|
| esv29993 | Chr.11:63971170 - 64096739 on Build GRCh38 | Loss |
|
| nsv555181 | Chr.11:63872409 - 64015880 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map