Assay Details
Target Gene Details
Entrez Gene ID: | 25834 |
Gene Name: | MGAT4 family member C |
Gene Aliases: |
GNTIVH, HGNT-IV-H |
Location: |
Chr.12:85972281-86838999 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT4C | NM_013244.3 | 7 | 1342 | NP_037376.2 |
| XM_005268776.4 | 6 | 1099 | XP_005268833.1 | |
| XM_005268779.3 | 5 | 1867 | XP_005268836.1 | |
| XM_011538150.2 | 6 | 1166 | XP_011536452.1 | |
| XM_011538151.2 | 6 | 1082 | XP_011536453.1 | |
| XM_011538156.2 | 9 | 2374 | XP_011536458.1 | |
| XM_011538157.2 | 8 | 3033 | XP_011536459.1 | |
| XM_017019143.1 | 6 | 1891 | XP_016874632.1 | |
| XM_017019144.1 | 6 | 1072 | XP_016874633.1 | |
| XM_017019145.1 | 7 | 1207 | XP_016874634.1 | |
| XM_017019146.1 | 7 | 1204 | XP_016874635.1 | |
| XM_017019147.1 | 6 | 1182 | XP_016874636.1 | |
| XM_017019148.1 | 8 | 2166 | XP_016874637.1 | |
| XM_017019149.1 | 7 | 2088 | XP_016874638.1 | |
| XM_017019150.1 | 8 | 1470 | XP_016874639.1 | |
| AK127727.1 | 8 | 2081 | ||
| AK299253.1 | 6 | 1099 | ||
| BC026068.1 | 6 | 1106 | AAH26068.1 | |
| BC064141.1 | 3 | 883 | AAH64141.1 | |
| BI667691.1 | 1 | 252 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049179 | Chr.12:85662598 - 86405410 on Build GRCh38 | Gain |
|
| nsv559667 | Chr.12:85924226 - 86022434 on Build GRCh38 | Loss |
|
| esv3892241 | Chr.12:85305039 - 88102633 on Build GRCh38 | Gain |
|
| nsv797 | Chr.12:85949715 - 85983671 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map