Assay Details
Target Gene Details
Entrez Gene ID: | 57337 |
Gene Name: | SUMO1/sentrin specific peptidase 7 |
Gene Aliases: |
- |
Location: |
Chr.3:101324189-101513241 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP7 | NM_001077203.2 | 23 | 3008 | NP_001070671.1 |
| NM_001282801.1 | 23 | 3005 | NP_001269730.1 | |
| NM_001282802.1 | 23 | 3153 | NP_001269731.1 | |
| NM_001282803.1 | 21 | 2760 | NP_001269732.1 | |
| NM_020654.4 | 24 | 3203 | NP_065705.3 | |
| XM_005247662.4 | 22 | 2796 | XP_005247719.1 | |
| XM_011513038.2 | 24 | 3192 | XP_011511340.1 | |
| XM_011513039.2 | 23 | 3117 | XP_011511341.1 | |
| XM_011513040.2 | 23 | 2997 | XP_011511342.1 | |
| XM_011513041.2 | 23 | 2994 | XP_011511343.1 | |
| XM_011513042.2 | 22 | 2799 | XP_011511344.1 | |
| XM_017006924.1 | 24 | 3080 | XP_016862413.1 | |
| XM_017006925.1 | 24 | 3168 | XP_016862414.1 | |
| XM_017006926.1 | 22 | 2906 | XP_016862415.1 | |
| XM_017006927.1 | 22 | 2903 | XP_016862416.1 | |
| AB051494.1 | 17 | 2210 | BAB21798.1 | |
| AF199458.1 | 23 | 3151 | AAL25651.1 | |
| AF217504.1 | 7 | 1019 | AAG09703.1 | |
| AL136599.1 | 23 | 3031 | CAB66534.1 | |
| AL833577.1 | 23 | 2974 | ||
| BC129988.1 | 24 | 3130 | ||
| BX537943.1 | 21 | 2704 | CAD97911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003475 | Chr.3:101286394 - 101401186 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map