Assay Details
Target Gene Details
Entrez Gene ID: | 57512 |
Gene Name: | G protein-coupled receptor 158 |
Gene Aliases: |
- |
Location: |
Chr.10:25174722-25602229 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR158 | NM_020752.2 | 1 | 121 | NP_065803.2 |
| AK095120.1 | 1 | 420 | ||
| AY528411.1 | 1 | 121 | AAS18315.1 |
Target Gene Details
Entrez Gene ID: | 100128811 |
Gene Name: | GPR158 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:25158072-25176276 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR158-AS1 | NR_027333.1 | |||
| AK094758.1 |
More Information
Set Membership: |
|

Genomic Map