Assay Details
Target Gene Details
Entrez Gene ID: | 84166 |
Gene Name: | NLR family CARD domain containing 5 |
Gene Aliases: |
CLR16.1, NOD27, NOD4 |
Location: |
Chr.16:56989547-57083524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059098 | Chr.16:56733938 - 57028336 on Build GRCh38 | Gain |
|
| nsv827683 | Chr.16:57016822 - 57147283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map