Assay Details
Target Gene Details
Entrez Gene ID: | 2047 |
Gene Name: | EPH receptor B1 |
Gene Aliases: |
ELK, EPHT2, Hek6, NET |
Location: |
Chr.3:134776089-135260467 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHB1 | NM_004441.4 | 1 | 300 | NP_004432.1 |
| XM_017005866.1 | 1 | 300 | XP_016861355.1 | |
| XM_017005868.1 | XP_016861357.1 | |||
| AK291208.1 | 1 | 300 | ||
| AK309741.1 | 1 | 297 | ||
| BC111744.1 | 1 | 96 | ||
| DA107464.1 | 1 | 301 | ||
| L40636.1 | 1 | 145 | AAB08520.1 |
More Information
Set Membership: |
|

Genomic Map