Assay Details
Target Gene Details
Entrez Gene ID: | 55502 |
Gene Name: | hes family bHLH transcription factor 6 |
Gene Aliases: |
C-HAIRY1, HES-6, bHLHb41, bHLHc23 |
Location: |
Chr.2:238238267-238240124 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HES6 | NM_001142853.2 | 4 | 1265 | NP_001136325.1 |
| NM_001282434.1 | 4 | 1231 | NP_001269363.1 | |
| NM_018645.5 | 4 | 1271 | NP_061115.2 | |
| AF260237.1 | 4 | 1112 | AAK51634.1 | |
| AK075040.1 | 3 | 1420 | BAC11368.1 | |
| BC007939.2 | 4 | 1176 | AAH07939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584761 | Chr.2:238212011 - 238246412 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
| dgv7349n54 | Chr.2:238238228 - 238241307 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map