Assay Details
Target Gene Details
Entrez Gene ID: | 81608 |
Gene Name: | factor interacting with PAPOLA and CPSF1 |
Gene Aliases: |
FIP1, Rhe, hFip1 |
Location: |
Chr.4:53377572-53462611 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIP1L1 | NM_001134937.1 | 1 | 57 | NP_001128409.1 |
| NM_001134938.1 | 1 | 57 | NP_001128410.1 | |
| NM_030917.3 | 1 | 57 | NP_112179.2 | |
| XM_005265769.4 | 1 | 124 | XP_005265826.1 | |
| XM_005265773.4 | 1 | 126 | XP_005265830.1 | |
| XM_005265774.4 | 1 | 65 | XP_005265831.1 | |
| XM_005265778.4 | 1 | 109 | XP_005265835.1 | |
| XM_005265779.4 | 1 | 64 | XP_005265836.1 | |
| XM_005265781.4 | 1 | 118 | XP_005265838.1 | |
| XM_005265782.4 | 1 | 64 | XP_005265839.1 | |
| XM_017008662.1 | 1 | 74 | XP_016864151.1 | |
| XM_017008663.1 | 1 | 66 | XP_016864152.1 | |
| XM_017008664.1 | 1 | 127 | XP_016864153.1 | |
| XM_017008665.1 | 1 | 64 | XP_016864154.1 | |
| XM_017008666.1 | 1 | 87 | XP_016864155.1 | |
| XM_017008667.1 | 1 | 65 | XP_016864156.1 | |
| XM_017008668.1 | 1 | 138 | XP_016864157.1 | |
| XM_017008669.1 | 1 | 80 | XP_016864158.1 | |
| XM_017008670.1 | 1 | 64 | XP_016864159.1 | |
| XM_017008671.1 | 1 | 64 | XP_016864160.1 | |
| AK295737.1 | 1 | 72 | ||
| AL136910.1 | CAB66844.1 | |||
| AY366510.1 | AAQ88277.1 | |||
| BC017724.1 | AAH17724.1 | |||
| BC110383.1 | 1 | 50 | ||
| DN990103.1 | 1 | 66 |
More Information
Set Membership: |
|

Genomic Map