Assay Details
Target Gene Details
Entrez Gene ID: | 114112 |
Gene Name: | thioredoxin reductase 3 |
Gene Aliases: |
TGR, TR2, TRXR3 |
Location: |
Chr.3:126607052-126655124 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNRD3 | NM_001173513.1 | 15 | 2121 | NP_001166984.1 |
| NM_052883.1 | 16 | 2229 | NP_443115.1 | |
| AF133519.1 | 16 | 1897 | AAD39929.1 | |
| AF171055.1 | 16 | 1903 | AAD51325.1 | |
| AK299502.1 | 15 | 2098 | ||
| BC030028.2 | 16 | 2207 | AAH30028.1 | |
| BC050032.1 | 16 | 2216 | AAH50032.1 |
Target Gene Details
Entrez Gene ID: | 645840 |
Gene Name: | thioredoxin reductase 3 neighbor |
Gene Aliases: |
TR2IT1, TXNRD3IT1, TXNRD3NT1 |
Location: |
Chr.3:126571779-126608555 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNRD3NB | NM_001039783.1 | 2 | 289 | NP_001034872.1 |
| AB113648.1 | 2 | 289 | BAD26752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4852n100 | Chr.3:126607261 - 126669480 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map