Assay Details
Target Gene Details
Entrez Gene ID: | 9113 |
Gene Name: | large tumor suppressor kinase 1 |
Gene Aliases: |
WARTS, wts |
Location: |
Chr.6:149658153-149718256 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LATS1 | NM_004690.3 | 8 | 4704 | NP_004681.1 |
| NR_073033.1 | 9 | 4610 | ||
| XM_017011474.1 | 9 | 4645 | XP_016866963.1 | |
| XM_017011475.1 | 9 | 4565 | XP_016866964.1 | |
| XM_017011476.1 | 10 | 4713 | XP_016866965.1 | |
| XM_017011478.1 | 9 | 4623 | XP_016866967.1 | |
| XM_017011480.1 | 8 | 4342 | XP_016866969.1 | |
| XM_017011481.1 | 7 | 4388 | XP_016866970.1 | |
| AF164041.1 | AAD50272.1 |
More Information
Set Membership: |
|

Genomic Map