Assay Details
Target Gene Details
Entrez Gene ID: | 11235 |
Gene Name: | programmed cell death 10 |
Gene Aliases: |
CCM3, TFAR15 |
Location: |
Chr.3:167683891-167735690 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDCD10 | NM_007217.3 | 9 | 1033 | NP_009148.2 |
| NM_145859.1 | 9 | 882 | NP_665858.1 | |
| NM_145860.1 | 8 | 788 | NP_665859.1 | |
| XM_005247086.4 | 9 | 1768 | XP_005247143.1 | |
| XM_005247087.4 | 9 | 1773 | XP_005247144.1 | |
| XM_005247088.3 | 8 | 864 | XP_005247145.1 | |
| XM_006713485.3 | 9 | 928 | XP_006713548.1 | |
| XM_011512368.2 | 10 | 1841 | XP_011510670.1 | |
| XM_011512369.2 | 10 | 1846 | XP_011510671.1 | |
| XM_017005644.1 | 9 | 923 | XP_016861133.1 | |
| XM_017005645.1 | 9 | 1764 | XP_016861134.1 | |
| AF022385.1 | 8 | 788 | AAB72225.1 | |
| AK291130.1 | 8 | 819 | ||
| BC002506.1 | 9 | 1045 | AAH02506.1 | |
| BC016353.1 | 9 | 886 | AAH16353.1 | |
| CR457107.1 | CAG33388.1 |
More Information
Set Membership: |
|

Genomic Map