Assay Details
Target Gene Details
Entrez Gene ID: | 9182 |
Gene Name: | Ras association domain family member 9 |
Gene Aliases: |
P-CIP1, PAMCI, PCIP1 |
Location: |
Chr.12:85800703-85836540 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF9 | NM_005447.3 | 2 | 524 | NP_005438.2 |
| XM_011538988.2 | 2 | 442 | XP_011537290.1 | |
| AF056209.1 | 2 | 302 | AAD03250.1 | |
| AK022089.1 | 2 | 365 | ||
| BC031589.1 | 2 | 316 | AAH31589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049179 | Chr.12:85662598 - 86405410 on Build GRCh38 | Gain |
|
| dgv1539n100 | Chr.12:85792109 - 85878715 on Build GRCh38 | Gain |
|
| esv3892242 | Chr.12:85514618 - 85885975 on Build GRCh38 | Loss |
|
| dgv1537n100 | Chr.12:85264824 - 85841655 on Build GRCh38 | Loss |
|
| esv3892241 | Chr.12:85305039 - 88102633 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map