Assay Details
Target Gene Details
Entrez Gene ID: | 27301 |
Gene Name: | apurinic/apyrimidinic endodeoxyribonuclease 2 |
Gene Aliases: |
APE2, APEXL2, XTH2, ZGRF2 |
Location: |
Chr.X:55000323-55007873 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APEX2 | NM_001271748.1 | NP_001258677.1 | ||
| NM_014481.3 | NP_055296.2 | |||
| AB021260.1 | BAA78422.1 | |||
| AB049211.1 | BAB13764.1 | |||
| AF119046.1 | AAD43041.1 | |||
| AJ011311.1 | CAB45242.1 | |||
| AK301555.1 | ||||
| AK316186.1 | ||||
| BC002959.1 | AAH02959.1 | |||
| BI837686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| esv3573984 | Chr.X:54997314 - 55007120 on Build GRCh38 | Loss |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map