Assay Details
Target Gene Details
Entrez Gene ID: | 57570 |
Gene Name: | tRNA methyltransferase 5 |
Gene Aliases: |
COXPD26, KIAA1393, TRM5 |
Location: |
Chr.14:60971441-60981170 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT5 | NM_020810.3 | 2 | 226 | NP_065861.3 |
| XM_005267916.3 | 2 | 256 | XP_005267973.2 | |
| XM_011537017.2 | 2 | 317 | XP_011535319.1 | |
| AB037814.1 | 1 | 95 | BAA92631.1 | |
| AK293158.1 | 2 | 196 | ||
| BC108284.1 | 2 | 197 | AAI08285.1 | |
| BC136606.1 | 2 | 141 | ||
| BC136607.1 | 2 | 141 | ||
| BQ050607.1 | 2 | 204 | ||
| DB448202.2 | 2 | 212 | ||
| DC370229.1 | 2 | 226 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037387 | Chr.14:60742756 - 61066784 on Build GRCh38 | Gain |
|
| esv2748755 | Chr.14:60887693 - 61016326 on Build GRCh38 | Deletion |
|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
| nsv1049936 | Chr.14:60934305 - 61080658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map