Assay Details
Target Gene Details
Entrez Gene ID: | 220002 |
Gene Name: | cytochrome b561 family member A3 |
Gene Aliases: |
CYBASC3, LCYTB |
Location: |
Chr.11:61348748-61362299 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB561A3 | NM_001161452.1 | 5 | 1825 | NP_001154924.1 |
| NM_001161454.1 | 8 | 2480 | NP_001154926.1 | |
| NM_001300763.1 | 7 | 1532 | NP_001287692.1 | |
| NM_153611.4 | 7 | 2311 | NP_705839.3 | |
| XM_011544821.1 | 8 | 1717 | XP_011543123.1 | |
| XM_011544823.1 | 6 | 1452 | XP_011543125.1 | |
| XM_017017345.1 | 9 | 3634 | XP_016872834.1 | |
| XM_017017346.1 | 8 | 3465 | XP_016872835.1 | |
| XM_017017347.1 | 6 | 1074 | XP_016872836.1 | |
| AK024953.1 | 4 | 1275 | ||
| AK025870.1 | 3 | 1158 | ||
| AK056751.1 | 7 | 2302 | ||
| AK075559.1 | 7 | 1912 | BAC11698.1 | |
| AL833374.1 | 3 | 1142 | ||
| BC004391.1 | 7 | 2285 | AAH04391.1 | |
| BC014045.2 | 7 | 2121 | AAH14045.2 | |
| BC047710.1 | 7 | 1830 | AAH47710.1 | |
| BM692535.1 | 1 | 332 | ||
| CF541093.1 | 1 | 690 |
Target Gene Details
Entrez Gene ID: | 26007 |
Gene Name: | triokinase and FMN cyclase |
Gene Aliases: |
DAK, NET45 |
Location: |
Chr.11:61333182-61353426 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TKFC | XM_017017518.1 | XP_016873007.1 | ||
| XM_017017522.1 | XP_016873011.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428259 | Chr.11:61049513 - 61356164 on Build GRCh38 | Gain+Loss |
|
| nsv832177 | Chr.11:61185118 - 61356164 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map