Assay Details
Target Gene Details
Entrez Gene ID: | 4858 |
Gene Name: | NOVA alternative splicing regulator 2 |
Gene Aliases: |
ANOVA, NOVA3 |
Location: |
Chr.19:45933734-45973399 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOVA2 | NM_002516.3 | 4 | 1481 | NP_002507.1 |
| XM_006723230.3 | 5 | 1533 | XP_006723293.1 | |
| XM_017026838.1 | 4 | 1757 | XP_016882327.1 | |
| XM_017026839.1 | 4 | 1583 | XP_016882328.1 | |
| XM_017026840.1 | 3 | 1340 | XP_016882329.1 | |
| AF083898.1 | 4 | 1481 | AAC72355.1 | |
| U70477.1 | 4 | 1452 | AAB88661.1 |
More Information
Set Membership: |
|

Genomic Map