Assay Details
Target Gene Details
Entrez Gene ID: | 775 |
Gene Name: | calcium voltage-gated channel subunit alpha1 C |
Gene Aliases: |
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS |
Location: |
Chr.12:1969677-2697949 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 196513 |
Gene Name: | decapping mRNA 1B |
Gene Aliases: |
DCP1 |
Location: |
Chr.12:1941591-2004535 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCP1B | NM_001319292.1 | NP_001306221.1 | ||
| NM_152640.4 | NP_689853.3 | |||
| NR_135060.1 | ||||
| XM_011520927.2 | XP_011519229.1 | |||
| AK056200.1 | BAB71118.1 | |||
| AK299203.1 | ||||
| AK301109.1 | ||||
| AK310475.1 | ||||
| AY146652.1 | AAN62764.1 | |||
| BC043437.2 | AAH43437.1 | |||
| BM544996.1 | ||||
| DC425587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521838 | Chr.12:1951414 - 2190514 on Build GRCh38 | Gain |
|
| nsv1042378 | Chr.12:1968731 - 2020607 on Build GRCh38 | Gain |
|
| nsv1040370 | Chr.12:1912229 - 2561851 on Build GRCh38 | Gain |
|
| nsv574 | Chr.12:2002711 - 2035810 on Build GRCh38 | Insertion |
|
| nsv525455 | Chr.12:1956204 - 2025860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map