Assay Details
Target Gene Details
Entrez Gene ID: | 2149 |
Gene Name: | coagulation factor II thrombin receptor |
Gene Aliases: |
CF2R, HTR, PAR-1, PAR1, TR |
Location: |
Chr.5:76716043-76735780 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2R | NM_001311313.1 | 3 | 808 | NP_001298242.1 |
| NM_001992.4 | 2 | 686 | NP_001983.2 | |
| AK297131.1 | 2 | 379 | ||
| AK308169.1 | 2 | 686 | ||
| AK314202.1 | 2 | 609 | ||
| BC002464.2 | 2 | 601 | AAH02464.1 | |
| BC051909.1 | 2 | 607 | AAH51909.1 | |
| BT007279.1 | 2 | 421 | AAP35943.1 | |
| M62424.1 | 2 | 645 | AAA36743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035135 | Chr.5:76631202 - 76796233 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map