Assay Details
Target Gene Details
Entrez Gene ID: | 83903 |
Gene Name: | germ cell associated 2, haspin |
Gene Aliases: |
HASPIN |
Location: |
Chr.17:3723903-3726699 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSG2 | NM_031965.2 | 1 | 2001 | NP_114171.2 |
| AB039834.3 | 1 | 1998 | ||
| AF289865.1 | 1 | 1997 | AAK30300.1 | |
| AK056691.1 | BAB71255.1 | |||
| AK292715.1 | 1 | 1987 | ||
| AK310546.1 | ||||
| AK315247.1 | 1 | 2001 | ||
| BC016626.1 | 1 | 1987 | ||
| BC047457.1 | 1 | 1982 | AAH47457.1 |
Target Gene Details
Entrez Gene ID: | 3682 |
Gene Name: | integrin subunit alpha E |
Gene Aliases: |
CD103, HUMINAE |
Location: |
Chr.17:3714625-3803766 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGAE | NM_002208.4 | NP_002199.3 | ||
| XM_011523823.2 | XP_011522125.1 | |||
| XM_011523825.2 | XP_011522127.1 | |||
| XM_011523827.2 | XP_011522129.1 | |||
| XM_011523828.2 | XP_011522130.1 | |||
| XM_017024586.1 | XP_016880075.1 | |||
| BC113436.1 | ||||
| BC117207.1 | ||||
| L25851.2 | AAB59359.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| nsv574237 | Chr.17:3486822 - 3729542 on Build GRCh38 | Loss |
|
| nsv469791 | Chr.17:3700760 - 3885899 on Build GRCh38 | Loss |
|
| nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain |
|
| nsv833342 | Chr.17:3626188 - 3806082 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map