Assay Details
Target Gene Details
Entrez Gene ID: | 4283 |
Gene Name: | C-X-C motif chemokine ligand 9 |
Gene Aliases: |
CMK, Humig, MIG, SCYB9, crg-10 |
Location: |
Chr.4:76001342-76007523 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL9 | NM_002416.2 | 4 | 993 | NP_002407.1 |
| BC063122.1 | 4 | 984 | AAH63122.1 | |
| BC095396.1 | 4 | 979 | AAH95396.1 | |
| X72755.1 | 4 | 958 | CAA51284.1 |
Target Gene Details
Entrez Gene ID: | 101928809 |
Gene Name: | uncharacterized LOC101928809 |
Gene Aliases: |
- |
Location: |
Chr.4:75980790-76005942 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928809 | NR_125906.1 |
More Information
Set Membership: |
|

Genomic Map