Assay Details
Target Gene Details
Entrez Gene ID: | 55697 |
Gene Name: | Vac14, PIKFYVE complex component |
Gene Aliases: |
ArPIKfyve, TAX1BP2, TRX |
Location: |
Chr.16:70687439-70801169 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAC14 | NM_018052.3 | NP_060522.3 | ||
| XM_005256038.3 | XP_005256095.1 | |||
| AK001167.1 | BAA91530.1 | |||
| AK025479.1 | BAB15145.1 | |||
| AK056433.1 | ||||
| AK093941.1 | ||||
| AK297564.1 | ||||
| AK307766.1 | ||||
| AK310466.1 | 4 | 592 | ||
| BC000536.2 | AAH00536.2 | |||
| BC007214.1 | AAH07214.2 | |||
| BC125108.1 | ||||
| BC125109.1 | ||||
| CR457343.1 | CAG33624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv572947 | Chr.16:70680531 - 70697967 on Build GRCh38 | Loss |
|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
| nsv572948 | Chr.16:70690529 - 70697967 on Build GRCh38 | Loss |
|
| nsv517391 | Chr.16:70632507 - 70806587 on Build GRCh38 | Loss |
|
| nsv572945 | Chr.16:70632819 - 70806587 on Build GRCh38 | Gain |
|
| nsv1059226 | Chr.16:70674795 - 70745241 on Build GRCh38 | Gain |
|
| nsv471094 | Chr.16:70660097 - 70716067 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map