Assay Details
Target Gene Details
Entrez Gene ID: | 167681 |
Gene Name: | protease, serine 35 |
Gene Aliases: |
C6orf158, dJ223E3.1 |
Location: |
Chr.6:83512475-83525704 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS35 | NM_001170423.1 | 3 | 977 | NP_001163894.1 |
| NM_153362.2 | 2 | 872 | NP_699193.2 | |
| AK291928.1 | 2 | 757 | ||
| AY358661.1 | 3 | 906 | AAQ89024.1 | |
| BC037170.1 | 2 | 795 | AAH37170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518229 | Chr.6:83519395 - 83524112 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map