Assay Details
Target Gene Details
Entrez Gene ID: | 100529261 |
Gene Name: | CHURC1-FNTB readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:64914361-65062655 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHURC1-FNTB | NM_001202558.1 | NP_001189487.1 | ||
| NM_001202559.1 | NP_001189488.1 | |||
| AK296850.1 | ||||
| AK303739.1 |
Target Gene Details
Entrez Gene ID: | 2342 |
Gene Name: | farnesyltransferase, CAAX box, beta |
Gene Aliases: |
FPTB |
Location: |
Chr.14:64986789-65062652 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNTB | NM_002028.3 | NP_002019.1 | ||
| AK093298.1 | ||||
| AK225917.1 | ||||
| AK295972.1 | ||||
| AK315714.1 | ||||
| AU122768.1 | ||||
| BC020232.1 | AAH20232.1 | |||
| BX248269.1 | CAD62597.1 | |||
| L00635.1 | AAA35854.1 | |||
| L10414.1 | AAA86286.1 |
Target Gene Details
Entrez Gene ID: | 4149 |
Gene Name: | MYC associated factor X |
Gene Aliases: |
bHLHd4 |
Location: |
Chr.14:65006101-65102695 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAX | NM_001271069.1 | NP_001257998.1 | ||
| NM_197957.3 | NP_932061.1 | |||
| AI652319.1 | ||||
| AI655674.1 | ||||
| AI917842.1 | ||||
| DB482510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832816 | Chr.14:65006526 - 65177927 on Build GRCh38 | Gain |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map