Assay Details
Target Gene Details
Entrez Gene ID: | 1889 |
Gene Name: | endothelin converting enzyme 1 |
Gene Aliases: |
ECE |
Location: |
Chr.1:21217247-21345541 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECE1 | NM_001113347.1 | 10 | 1657 | NP_001106818.1 |
| NM_001113348.1 | 12 | 1552 | NP_001106819.1 | |
| NM_001113349.1 | 11 | 1503 | NP_001106820.1 | |
| NM_001397.2 | 12 | 1512 | NP_001388.1 | |
| XM_006710398.2 | 12 | 1657 | XP_006710461.1 | |
| XM_011540872.2 | 12 | 2264 | XP_011539174.1 | |
| XM_011540873.2 | 12 | 1582 | XP_011539175.1 | |
| XM_017000511.1 | 12 | 1404 | XP_016856000.1 | |
| AB031742.1 | 11 | 1397 | BAA83687.1 | |
| AK290656.1 | 12 | 1484 | ||
| AK296486.1 | 12 | 1487 | ||
| AK304167.1 | 11 | 1445 | ||
| AK309677.1 | 12 | 1486 | ||
| BC117256.1 | 12 | 1512 | ||
| BC126257.1 | 12 | 1512 | ||
| BX648351.1 | 10 | 1516 | ||
| CN298523.1 | 4 | 409 | ||
| CN298525.1 | 3 | 194 | ||
| D43698.1 | 10 | 1462 | BAA07800.1 | |
| D49471.1 | 12 | 1437 | BAA08442.1 | |
| Z35307.1 | 12 | 1423 | CAA84548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834380 | Chr.1:21072777 - 21247948 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map