Assay Details
Target Gene Details
Entrez Gene ID: | 55624 |
Gene Name: | protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) |
Gene Aliases: |
GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2 |
Location: |
Chr.1:46188681-46220305 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMGNT1 | NM_001243766.1 | NP_001230695.1 | ||
| NM_001290129.1 | NP_001277058.1 | |||
| NM_001290130.1 | NP_001277059.1 | |||
| NM_017739.3 | NP_060209.3 | |||
| XM_005271010.1 | XP_005271067.1 | |||
| XM_006710755.1 | XP_006710818.1 | |||
| XM_006710756.1 | XP_006710819.1 | |||
| XM_011541760.2 | XP_011540062.1 | |||
| XM_017001690.1 | XP_016857179.1 | |||
| AB057356.1 | BAB71960.1 | |||
| AF250859.2 | AAF71270.2 | |||
| AK000284.1 | BAA91053.1 | |||
| AK022727.1 | BAB14207.1 | |||
| AK056186.1 | ||||
| AK225033.1 | ||||
| AK301923.1 | ||||
| AK302379.1 | ||||
| AK316399.1 | ||||
| AY358592.1 | AAQ88955.1 | |||
| BC001471.2 | AAH01471.1 | |||
| CR749700.1 | CAH18478.2 |
Target Gene Details
Entrez Gene ID: | 10103 |
Gene Name: | tetraspanin 1 |
Gene Aliases: |
NET1, TM4C, TM4SF |
Location: |
Chr.1:46175077-46196489 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN1 | XM_011540460.2 | XP_011538762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470711 | Chr.1:46194623 - 46344858 on Build GRCh38 | Gain |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
| nsv822520 | Chr.1:46184814 - 46196029 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map