Assay Details
Target Gene Details
Entrez Gene ID: | 84838 |
Gene Name: | zinc finger protein 496 |
Gene Aliases: |
NIZP1, ZFP496, ZKSCAN17, ZSCAN49 |
Location: |
Chr.1:247297413-247333376 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF496 | NM_032752.1 | 9 | 1819 | NP_116141.1 |
| XM_005273328.3 | 11 | 3418 | XP_005273385.1 | |
| XM_005273330.3 | 10 | 3310 | XP_005273387.1 | |
| XM_011544310.1 | 12 | 3511 | XP_011542612.1 | |
| XM_011544312.1 | 11 | 3403 | XP_011542614.1 | |
| XM_017002586.1 | 13 | 3604 | XP_016858075.1 | |
| XM_017002587.1 | 13 | 3557 | XP_016858076.1 | |
| XM_017002588.1 | 12 | 3511 | XP_016858077.1 | |
| XM_017002589.1 | 12 | 3496 | XP_016858078.1 | |
| XM_017002590.1 | 11 | 3403 | XP_016858079.1 | |
| XM_017002591.1 | 10 | 3263 | XP_016858080.1 | |
| XM_017002594.1 | 6 | 851 | XP_016858083.1 | |
| AK024643.1 | 1 | 308 | ||
| BC007263.1 | 9 | 1819 | AAH07263.1 | |
| BC025794.1 | 8 | 1129 | AAH25794.1 | |
| BX647480.1 | 2 | 5754 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833126 | Chr.1:247109782 - 247312848 on Build GRCh38 | Loss |
|
| nsv833137 | Chr.1:247274015 - 247444126 on Build GRCh38 | Gain |
|
| esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss |
|
| esv3589514 | Chr.1:247297392 - 247354590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map