Assay Details
Target Gene Details
Entrez Gene ID: | 9896 |
Gene Name: | FIG4 phosphoinositide 5-phosphatase |
Gene Aliases: |
ALS11, BTOP, CMT4J, KIAA0274, SAC3, YVS, dJ249I4.1 |
Location: |
Chr.6:109691216-109825431 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIG4 | NM_014845.5 | 1 | 114 | NP_055660.1 |
| XM_017011591.1 | 1 | 119 | XP_016867080.1 | |
| XM_017011592.1 | XP_016867081.1 | |||
| XM_017011593.1 | XP_016867082.1 | |||
| AK222732.1 | BAD96452.1 | |||
| AK291671.1 | ||||
| AK293422.1 | ||||
| BC041338.2 | 1 | 70 | AAH41338.1 | |
| D87464.1 | ||||
| DA811740.1 | 1 | 119 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830769 | Chr.6:109623131 - 109756917 on Build GRCh38 | Gain |
|
| nsv604485 | Chr.6:109638066 - 109733283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map