Assay Details
Target Gene Details
Entrez Gene ID: | 51816 |
Gene Name: | cat eye syndrome chromosome region, candidate 1 |
Gene Aliases: |
ADA2, ADGF, IDGFL, PAN, SNEDS |
Location: |
Chr.22:17178790-17221854 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR1 | NM_001282225.1 | 10 | 1855 | NP_001269154.1 |
| NM_001282226.1 | 10 | 1822 | NP_001269155.1 | |
| NM_001282227.1 | 10 | 1628 | NP_001269156.1 | |
| NM_001282228.1 | 10 | 1730 | NP_001269157.1 | |
| NM_001282229.1 | 9 | 1487 | NP_001269158.1 | |
| NM_177405.2 | 7 | 1061 | NP_803124.1 | |
| XM_006724080.3 | 9 | 3092 | XP_006724143.1 | |
| XM_011546133.1 | 10 | 1747 | XP_011544435.1 | |
| AB209648.1 | 9 | 3092 | BAD92885.1 | |
| AF190746.1 | 9 | 1791 | AAF65941.1 | |
| AK027682.1 | 7 | 882 | BAB55293.1 | |
| AK074702.1 | 7 | 933 | BAC11148.1 | |
| AK130725.1 | 4 | 575 | ||
| AK292689.1 | 10 | 1843 | ||
| AK304818.1 | 10 | 1622 | ||
| AK314321.1 | ||||
| BC051755.1 | 9 | 1626 | AAH51755.1 | |
| CR456417.1 | 9 | 1613 | CAG30303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647209 | Chr.22:17151829 - 17264992 on Build GRCh38 | Gain |
|
| nsv1067241 | Chr.22:16757380 - 17351252 on Build GRCh38 | Loss |
|
| esv3647210 | Chr.22:17174250 - 17207794 on Build GRCh38 | Gain |
|
| nsv509805 | Chr.22:17081417 - 17267685 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map