Assay Details
Target Gene Details
Entrez Gene ID: | 2267 |
Gene Name: | fibrinogen like 1 |
Gene Aliases: |
HFREP1, HP-041, LFIRE-1, LFIRE1 |
Location: |
Chr.8:17864380-17909983 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGL1 | NM_004467.3 | 3 | 292 | NP_004458.3 |
| NM_147203.2 | 3 | 380 | NP_671736.2 | |
| NM_201552.1 | 4 | 419 | NP_963846.1 | |
| NM_201553.1 | 4 | 507 | NP_963847.1 | |
| XM_017013232.1 | 4 | 1269 | XP_016868721.1 | |
| XM_017013233.1 | 2 | 344 | XP_016868722.1 | |
| AF042354.1 | 3 | 327 | ||
| AF168954.1 | 3 | 315 | AAM88911.1 | |
| AK225616.1 | 3 | 278 | ||
| AK225619.1 | 3 | 267 | ||
| AK289448.1 | 3 | 278 | ||
| AL531132.3 | 3 | 658 | ||
| AX772838.1 | 4 | 372 | CAE06268.1 | |
| BC007047.1 | 3 | 288 | AAH07047.1 | |
| BG565379.1 | 4 | 445 | ||
| BG566131.1 | 4 | 402 | ||
| BG567956.1 | 3 | 381 | ||
| BG618053.1 | 3 | 293 | ||
| BT006635.1 | 2 | 183 | AAP35281.1 | |
| D14446.1 | 3 | 261 | BAA03336.1 | |
| D87342.1 | 2 | 188 | BAB70690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831250 | Chr.8:17737238 - 17890332 on Build GRCh38 | Loss |
|
| dgv7121n100 | Chr.8:17766079 - 17957752 on Build GRCh38 | Gain |
|
| nsv1034118 | Chr.8:17876103 - 17916258 on Build GRCh38 | Loss |
|
| nsv610701 | Chr.8:17787519 - 17956578 on Build GRCh38 | Gain |
|
| esv3616503 | Chr.8:17877278 - 17897727 on Build GRCh38 | Gain |
|
| esv3616500 | Chr.8:17771011 - 17959017 on Build GRCh38 | Gain |
|
| esv2762735 | Chr.8:17723646 - 17957764 on Build GRCh38 | Gain |
|
| dgv12044n54 | Chr.8:17868408 - 17896525 on Build GRCh38 | Gain |
|
| nsv610706 | Chr.8:17880740 - 17882183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map