Assay Details
Target Gene Details
Entrez Gene ID: | 1659 |
Gene Name: | DEAH-box helicase 8 |
Gene Aliases: |
DDX8, HRH1, PRP22, PRPF22 |
Location: |
Chr.17:43483865-43544787 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX8 | NM_001322219.1 | NP_001309148.1 |
Target Gene Details
Entrez Gene ID: | 2118 |
Gene Name: | ETS variant 4 |
Gene Aliases: |
E1A-F, E1AF, PEA3, PEAS3 |
Location: |
Chr.17:43527843-43546432 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETV4 | NM_001079675.2 | NP_001073143.1 | ||
| NM_001261437.1 | NP_001248366.1 | |||
| NM_001261438.1 | NP_001248367.1 | |||
| NM_001261439.1 | 1 | 283 | NP_001248368.1 | |
| NM_001986.2 | NP_001977.1 | |||
| XM_011524514.1 | XP_011522816.1 | |||
| XM_017024349.1 | XP_016879838.1 | |||
| AK290429.1 | ||||
| AK298872.1 | ||||
| AK299019.1 | ||||
| AK312934.1 | ||||
| AK315961.1 | ||||
| BC016623.1 | AAH16623.1 | |||
| BF304101.1 | 1 | 283 | ||
| D12765.1 | BAA02234.1 | |||
| U18018.1 | AAA95991.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761959 | Chr.17:43322294 - 43565935 on Build GRCh38 | Gain |
|
| nsv833453 | Chr.17:43431629 - 43612561 on Build GRCh38 | Gain+Loss |
|
| nsv470588 | Chr.17:43346885 - 43616240 on Build GRCh38 | Gain |
|
| esv3640630 | Chr.17:43500927 - 43550600 on Build GRCh38 | Gain |
|
| esv3640625 | Chr.17:43361371 - 43577834 on Build GRCh38 | Gain |
|
| nsv1065802 | Chr.17:43338154 - 43584333 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map