Assay Details
Target Gene Details
Entrez Gene ID: | 80146 |
Gene Name: | UDP-glucuronate decarboxylase 1 |
Gene Aliases: |
SDR6E1, UGD |
Location: |
Chr.2:106093303-106194339 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UXS1 | NM_001253875.1 | 15 | 1339 | NP_001240804.1 |
| NM_001253876.1 | 10 | 1476 | NP_001240805.1 | |
| NM_025076.4 | 15 | 1324 | NP_079352.2 | |
| NR_045607.1 | 14 | 1275 | ||
| XM_006712775.3 | 14 | 1197 | XP_006712838.1 | |
| XM_011511902.2 | 15 | 1318 | XP_011510204.2 | |
| XM_011511903.2 | 15 | 1301 | XP_011510205.2 | |
| XM_011511904.2 | 12 | 1646 | XP_011510206.1 | |
| XM_017005008.1 | 13 | 1139 | XP_016860497.1 | |
| XM_017005009.1 | 13 | 1124 | XP_016860498.1 | |
| XM_017005010.1 | 14 | 1182 | XP_016860499.1 | |
| XM_017005011.1 | 15 | 1426 | XP_016860500.1 | |
| XM_017005012.1 | 15 | 1361 | XP_016860501.1 | |
| XM_017005013.1 | 13 | 1884 | XP_016860502.1 | |
| XM_017005014.1 | 14 | 1102 | XP_016860503.1 | |
| AK027244.1 | 10 | 1476 | BAB15705.1 | |
| AK056022.1 | 4 | 2153 | ||
| AK075120.1 | 15 | 1286 | BAC11415.1 | |
| AK075170.1 | 15 | 1301 | BAC11448.1 | |
| AK122696.1 | 14 | 1240 | ||
| AK225592.1 | 10 | 1476 | ||
| AK290668.1 | 15 | 1299 | ||
| AL834439.1 | 2 | 1088 | CAD39099.2 | |
| AY147934.1 | 15 | 1291 | AAN39844.1 | |
| AY358541.1 | 15 | 1275 | AAQ88905.1 | |
| BC009819.2 | 15 | 1265 | AAH09819.2 | |
| BX366726.2 | 1 | 737 |
More Information
Set Membership: |
|

Genomic Map