Assay Details
Target Gene Details
Entrez Gene ID: | 4145 |
Gene Name: | megakaryocyte-associated tyrosine kinase |
Gene Aliases: |
CHK, CTK, HHYLTK, HYL, HYLTK, Lsk |
Location: |
Chr.19:3777969-3801812 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MATK | NM_002378.3 | 4 | 557 | NP_002369.2 |
| NM_139354.2 | 3 | 393 | NP_647611.1 | |
| NM_139355.2 | 4 | 616 | NP_647612.1 | |
| XM_011528018.1 | 4 | 771 | XP_011526320.1 | |
| AB593100.1 | 4 | 518 | ||
| AK055395.1 | 4 | 557 | ||
| AK225834.1 | 3 | 216 | ||
| AK315646.1 | 4 | 509 | ||
| AL137754.1 | 3 | 266 | CAB70906.2 | |
| BC000114.1 | 4 | 615 | AAH00114.1 | |
| BC003109.1 | 4 | 615 | AAH03109.1 | |
| BG396388.1 | 4 | 616 | ||
| BM925586.1 | 4 | 601 | ||
| BQ109562.1 | 3 | 325 | ||
| L18974.1 | 4 | 478 | AAA16703.1 | |
| S71669.1 | 3 | 217 | AAB30995.1 | |
| X77278.1 | 4 | 423 | CAA54493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509718 | Chr.19:3676947 - 3844683 on Build GRCh38 | Insertion |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map