Assay Details
Target Gene Details
Entrez Gene ID: | 10202 |
Gene Name: | dehydrogenase/reductase 2 |
Gene Aliases: |
HEP27, SDR25C1 |
Location: |
Chr.14:23630115-23645639 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS2 | NM_001318835.1 | NP_001305764.1 | ||
| NM_005794.3 | NP_005785.1 | |||
| NM_182908.4 | NP_878912.1 | |||
| XM_005267249.1 | XP_005267306.1 | |||
| XM_006720001.3 | XP_006720064.1 | |||
| XM_011536338.1 | XP_011534640.1 | |||
| XM_011536339.2 | XP_011534641.1 | |||
| AB209653.1 | 1 | 818 | BAD92890.1 | |
| AK222857.1 | BAD96577.1 | |||
| AK309945.1 | ||||
| AK309975.1 | ||||
| BC002786.2 | AAH02786.1 | |||
| U31875.1 | AAA82048.1 |
More Information
Set Membership: |
|

Genomic Map