Assay Details
Target Gene Details
Entrez Gene ID: | 29118 |
Gene Name: | DEAD-box helicase 25 |
Gene Aliases: |
GRTH |
Location: |
Chr.11:125903275-125923110 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX25 | NM_013264.4 | 12 | 1655 | NP_037396.3 |
| XM_011542790.2 | 12 | 1559 | XP_011541092.1 | |
| XM_011542791.2 | 12 | 1607 | XP_011541093.1 | |
| XM_017017626.1 | 12 | 1596 | XP_016873115.1 | |
| AF155140.2 | 12 | 1549 | AAF21371.2 | |
| BC035388.1 | 12 | 1501 | ||
| BC050360.1 | 12 | 1525 | ||
| CD657404.1 | 4 | 600 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv535 | Chr.11:125918491 - 125963371 on Build GRCh38 | Deletion |
|
| nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain |
|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map