Assay Details
Target Gene Details
Entrez Gene ID: | 53842 |
Gene Name: | claudin 22 |
Gene Aliases: |
CLDN21 |
Location: |
Chr.4:183318067-183320774 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLDN22 | NM_001111319.1 | 1 | 2198 | NP_001104789.1 |
| AK098064.1 | 1 | 1592 |
Target Gene Details
Entrez Gene ID: | 80014 |
Gene Name: | WW and C2 domain containing 2 |
Gene Aliases: |
BOMB |
Location: |
Chr.4:183098677-183320777 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2 | NM_024949.5 | 23 | 6609 | NP_079225.5 |
| XM_011532269.2 | XP_011530571.1 | |||
| AK091401.1 | 1 | 384 | ||
| AL832424.1 | 3 | 3234 | CAH10641.1 | |
| BX647378.1 | 21 | 6200 | CAH10569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030766 | Chr.4:183305034 - 183948875 on Build GRCh38 | Gain |
|
| nsv1029647 | Chr.4:183268304 - 183372226 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map