Assay Details
Target Gene Details
Entrez Gene ID: | 89886 |
Gene Name: | SLAM family member 9 |
Gene Aliases: |
CD2F-10, CD2F10, CD84-H1, CD84H1, SF2001 |
Location: |
Chr.1:159951492-159984730 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLAMF9 | NM_001146172.1 | NP_001139644.1 | ||
| NM_001146173.1 | NP_001139645.1 | |||
| NM_033438.3 | NP_254273.2 | |||
| XM_017002756.1 | XP_016858245.1 | |||
| AF275725.1 | AAK69052.1 | |||
| AY034613.1 | AAK61389.1 | |||
| AY358801.1 | AAQ89161.1 | |||
| BC070213.1 | AAH70213.1 | |||
| BC074754.2 | AAH74754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548055 | Chr.1:159948628 - 159970210 on Build GRCh38 | Loss |
|
| esv3578219 | Chr.1:159948904 - 159963437 on Build GRCh38 | Loss |
|
| esv3587705 | Chr.1:159952544 - 159964328 on Build GRCh38 | Loss |
|
| nsv520773 | Chr.1:159953169 - 159965779 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map