Assay Details
Target Gene Details
Entrez Gene ID: | 112611 |
Gene Name: | RWD domain containing 2A |
Gene Aliases: |
RWDD2, dJ747H23.2 |
Location: |
Chr.6:83193313-83198935 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RWDD2A | NM_001322335.1 | 3 | 927 | NP_001309264.1 |
| NM_001322336.1 | 4 | 1238 | NP_001309265.1 | |
| NM_001322337.1 | 4 | 1223 | NP_001309266.1 | |
| NM_001322339.1 | 4 | 1019 | NP_001309268.1 | |
| NM_033411.4 | 3 | 1146 | NP_219479.2 | |
| XM_017010222.1 | 3 | 1729 | XP_016865711.1 | |
| XM_017010223.1 | 3 | 1504 | XP_016865712.1 | |
| AK295724.1 | 2 | 1797 | ||
| AK302978.1 | 3 | 856 | ||
| AK313083.1 | ||||
| AW023870.1 | ||||
| BC010930.2 | 2 | 1015 | AAH10930.1 | |
| BC104816.1 | 2 | 931 | AAI04817.1 | |
| BC112003.1 | 2 | 931 | AAI12004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map