Assay Details
Target Gene Details
Entrez Gene ID: | 30814 |
Gene Name: | phospholipase A2 group IIE |
Gene Aliases: |
GIIE sPLA2, sPLA2-IIE |
Location: |
Chr.1:19920307-19923617 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2E | NM_014589.2 | NP_055404.1 | ||
| AF189279.1 | AAF36541.1 | |||
| BC069116.1 | AAH69116.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545730 | Chr.1:19910238 - 19924306 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map