Assay Details
Target Gene Details
Entrez Gene ID: | 55090 |
Gene Name: | mediator complex subunit 9 |
Gene Aliases: |
MED25 |
Location: |
Chr.17:17476986-17493220 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED9 | NM_018019.2 | 2 | 1320 | NP_060489.1 |
| AK001055.1 | 2 | 1320 | BAA91483.1 |
More Information
Set Membership: |
|

Genomic Map