Assay Details
Target Gene Details
Entrez Gene ID: | 84135 |
Gene Name: | UTP15, small subunit processome component |
Gene Aliases: |
NET21 |
Location: |
Chr.5:73565741-73583377 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTP15 | NM_001284430.1 | NP_001271359.1 | ||
| NM_001284431.1 | NP_001271360.1 | |||
| NM_032175.3 | NP_115551.2 | |||
| XM_011543680.2 | XP_011541982.1 | |||
| AK074217.1 | BAB85020.1 | |||
| AK300525.1 | ||||
| AK302021.1 | ||||
| AL831972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830346 | Chr.5:73503223 - 73642429 on Build GRCh38 | Loss |
|
| nsv830347 | Chr.5:73517327 - 73706624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map