Assay Details
Target Gene Details
Entrez Gene ID: | 51380 |
Gene Name: | cysteine sulfinic acid decarboxylase |
Gene Aliases: |
CSD, PCAP |
Location: |
Chr.12:53157663-53180909 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSAD | NM_001244705.1 | NP_001231634.1 | ||
| NM_015989.4 | NP_057073.4 | |||
| XM_011538442.2 | XP_011536744.2 | |||
| XM_011538446.2 | XP_011536748.1 | |||
| XM_011538449.1 | XP_011536751.1 | |||
| XM_011538451.1 | XP_011536753.1 | |||
| XM_017019416.1 | XP_016874905.1 | |||
| XM_017019417.1 | XP_016874906.1 | |||
| XM_017019418.1 | XP_016874907.1 | |||
| XM_017019419.1 | XP_016874908.1 | |||
| AF116545.1 | AAD32543.1 | |||
| AF116546.1 | AAD32544.1 | |||
| AF116547.1 | AAD32545.1 | |||
| AF116548.1 | AAD32546.1 | |||
| AK122681.1 | ||||
| AK128796.1 | ||||
| AK289659.1 | ||||
| AK296885.1 | 6 | 1089 | ||
| AK315265.1 | ||||
| BC052249.1 | AAH52249.1 | |||
| BC098278.1 | AAH98278.1 | |||
| BC098342.1 | AAH98342.1 | |||
| BC099717.1 | AAH99717.1 | |||
| BC105918.1 | AAI05919.1 | |||
| DA687716.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain |
|
| nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss |
|
| nsv470295 | Chr.12:53099603 - 53227927 on Build GRCh38 | Gain |
|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
| dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss |
|
| nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map