Assay Details
Target Gene Details
Entrez Gene ID: | 5530 |
Gene Name: | protein phosphatase 3 catalytic subunit alpha |
Gene Aliases: |
CALN, CALNA, CALNA1, CCN1, CNA1, PPP2B |
Location: |
Chr.4:101023430-101347471 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP3CA | NM_000944.4 | 14 | 4538 | NP_000935.1 |
| NM_001130691.1 | 13 | 4508 | NP_001124163.1 | |
| NM_001130692.1 | 12 | 4382 | NP_001124164.1 | |
| XM_017008365.1 | 14 | 3842 | XP_016863854.1 | |
| AL353950.1 | 14 | 4269 | CAB89253.1 | |
| AW973358.1 | 1 | 139 | ||
| EU192652.1 | 12 | 4382 | ||
| EU192653.1 | 8 | 3855 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601585 | Chr.4:100952589 - 101052381 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map