Assay Details
Target Gene Details
Entrez Gene ID: | 5562 |
Gene Name: | protein kinase AMP-activated catalytic subunit alpha 1 |
Gene Aliases: |
AMPK, AMPKa1 |
Location: |
Chr.5:40759379-40798195 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAA1 | NM_006251.5 | 9 | 1608 | NP_006242.5 |
| NM_206907.3 | 10 | 1653 | NP_996790.3 | |
| XM_006714482.1 | 8 | 1502 | XP_006714545.1 | |
| XM_017009624.1 | 10 | 2564 | XP_016865113.1 | |
| XM_017009625.1 | 9 | 2509 | XP_016865114.1 | |
| AB022017.1 | 9 | 1598 | BAA36547.1 | |
| AF100763.1 | 9 | 1578 | AAD43027.1 | |
| AK307546.1 | 10 | 1635 | ||
| AK312947.1 | 9 | 1590 | ||
| BC037303.2 | 9 | 1590 | AAH37303.1 | |
| BC048980.1 | 10 | 1832 | AAH48980.1 | |
| Y12856.1 | 3 | 696 | CAA73361.1 |
More Information
Set Membership: |
|

Genomic Map