Assay Details
Target Gene Details
Entrez Gene ID: | 5738 |
Gene Name: | prostaglandin F2 receptor inhibitor |
Gene Aliases: |
CD315, CD9P-1, EWI-F, FPRP, SMAP-6 |
Location: |
Chr.1:116909923-116990358 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGFRN | NM_020440.3 | 2 | 604 | NP_065173.2 |
| XM_017001874.1 | 2 | 375 | XP_016857363.1 | |
| AB037857.1 | 2 | 460 | ||
| AK074637.1 | 2 | 470 | BAC11104.1 | |
| BC098142.1 | AAH98142.1 | |||
| BC114520.1 | AAI14521.1 | |||
| BC139718.1 | ||||
| BC152454.1 | 2 | 460 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831159 | Chr.1:116844705 - 117040990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map